missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzaldehyde, 98+%

Product Code. 10444785 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass Bottle
Unit Size:
10mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10444785 250 mL Glass Bottle 250mL
10477110 10 mL Glass Bottle 10mL
10245440 50 mL Glass Bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10444785 Supplier Thermo Scientific Acros Supplier No. 119332500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 456-48-4 | C7H5FO | 124.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 456-48-4
Molecular Formula C7H5FO
Molecular Weight (g/mol) 124.11
MDL Number MFCD00003348
InChI Key PIKNVEVCWAAOMJ-UHFFFAOYSA-N
Synonym m-fluorobenzaldehyde, benzaldehyde, 3-fluoro, 3'-fluoro-benzaldehyde, 3-fluoro-benzaldehyde, meta-fluorobenzaldehyde, benzaldehyde, m-fluoro, 3-fc6h4cho, 3-fluorobenzaldehyde, 3-fluoro benzaldehyde, pubchem3477
PubChem CID 68009
IUPAC Name 3-fluorobenzaldehyde
SMILES C1=CC(=CC(=C1)F)C=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1700g/mL
Boiling Point 66°C to 68°C (20.0 mmHg)
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass Bottle
Linear Formula FC6H4CHO
Refractive Index 1.5170 to 1.519
Quantity 250 mL
Beilstein 07, II, 177
Specific Gravity 1.17
Formula Weight 124.11
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 3-Fluorobenzaldehyde
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Fluorobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.