missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzaldehyde, 97%

Product Code. 11480223
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480223 250 g 250g
11460223 10 g 10g
11470223 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480223 Supplier Thermo Scientific Alfa Aesar Supplier No. A11396.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 456-48-4 | C7H5FO | 124.114 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 456-48-4
Molecular Formula C7H5FO
Molecular Weight (g/mol) 124.114
MDL Number MFCD00003348
InChI Key PIKNVEVCWAAOMJ-UHFFFAOYSA-N
Synonym m-fluorobenzaldehyde, benzaldehyde, 3-fluoro, 3'-fluoro-benzaldehyde, 3-fluoro-benzaldehyde, meta-fluorobenzaldehyde, benzaldehyde, m-fluoro, 3-fc6h4cho, 3-fluorobenzaldehyde, 3-fluoro benzaldehyde, pubchem3477
PubChem CID 68009
IUPAC Name 3-fluorobenzaldehyde
SMILES C1=CC(=CC(=C1)F)C=O

Specifications

Density 1.174
Boiling Point 173°C
Flash Point 56°C (132°F)
Refractive Index 1.519
Quantity 250 g
UN Number UN1989
Beilstein 970178
Sensitivity Air Sensitive
Formula Weight 124.11
Percent Purity 97%
Chemical Name or Material 3-Fluorobenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.