missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoroanisole, 99%

Product Code. 11444333
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
250 g
Unit Size:
10g
250g
25g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444333 10 g 10g
11349489 25 g 25g
11454333 50 g 50g
11464333 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11444333 Supplier Thermo Scientific Alfa Aesar Supplier No. A10931.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 456-49-5 | C7H7FO | 126.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 456-49-5
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00000335
InChI Key MFJNOXOAIFNSBX-UHFFFAOYSA-N
Synonym 3-fluoroanisole, m-fluoroanisole, benzene, 1-fluoro-3-methoxy, 1-fluoro-3-methoxy-benzene, 2-fluoro-4-methoxybenzene, fr co1 wln, 3-fluoroanisol, 3-fluoro-anisole, 3-fluoro anisole, anisole, m-fluoro
PubChem CID 9975
IUPAC Name 1-fluoro-3-methoxybenzene
SMILES COC1=CC=CC(F)=C1

Specifications

Melting Point -35°C
Density 1.104
Boiling Point 158°C to 159°C
Flash Point 43°C (109°F)
Refractive Index 1.489
Quantity 10 g
UN Number UN1993
Beilstein 1858895
Formula Weight 126.13
Percent Purity 99%
Chemical Name or Material 3-Fluoroanisole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.