missing translation for 'onlineSavingsMsg'
Learn More

3'-Fluoroacetophenone, 99%, Thermo Scientific™

Product Code. 10636242
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 455-36-7
Molecular Formula C8H7FO
Molecular Weight (g/mol) 138.14
MDL Number MFCD00000338
InChI Key HCEKGPAHZCYRBZ-UHFFFAOYSA-N
Synonym 3'-fluoroacetophenone, 1-3-fluorophenyl ethanone, 3-fluoroacetophenone, m-fluoroacetophenone, ethanone, 1-3-fluorophenyl, 1-3-fluorophenyl ethan-1-one, acetophenone, 3'-fluoro, 3'-fluoro-acetophenone, 1-acetyl-3-fluorobenzene
PubChem CID 9967
SMILES CC(=O)C1=CC=CC(F)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1200g/mL
Boiling Point 81.0°C (9.0 mmHg)
Flash Point 80°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4COCH3
Refractive Index 1.5080 to 1.5100
Quantity 1 g
Beilstein 07, III, 961
Specific Gravity 1.12
IUPAC Name 1-(3-fluorophenyl)ethanone
Formula Weight 138.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3′-Fluoroacetophenone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'