missing translation for 'onlineSavingsMsg'
Learn More

3'-Fluoroacetophenone, 97%

Product Code. 11409263
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409263 25 g 25g
11499253 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11409263 Supplier Thermo Scientific Alfa Aesar Supplier No. A11733.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 455-36-7 | C8H7FO | 138.14 g/mol

3'-Fluoroacetophenone is used as raw material and intermediate in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′-Fluoroacetophenone is used as raw material and intermediate in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong bases, strong oxidizing agents, strong reducing agents. Ensure adequate ventilation during use. Use only in a chemical fume hood.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 455-36-7
Molecular Formula C8H7FO
Molecular Weight (g/mol) 138.14
MDL Number MFCD00000338
InChI Key HCEKGPAHZCYRBZ-UHFFFAOYSA-N
Synonym 3'-fluoroacetophenone, 1-3-fluorophenyl ethanone, 3-fluoroacetophenone, m-fluoroacetophenone, ethanone, 1-3-fluorophenyl, 1-3-fluorophenyl ethan-1-one, acetophenone, 3'-fluoro, 3'-fluoro-acetophenone, 1-acetyl-3-fluorobenzene
PubChem CID 9967
IUPAC Name 1-(3-fluorophenyl)ethanone
SMILES CC(=O)C1=CC=CC(F)=C1

Specifications

Melting Point -3°C
Density 1.13
Boiling Point 80°C to 81°C (9 mmHg)
Flash Point 80°C (176°F)
Refractive Index 1.509
Quantity 25 g
Beilstein 907041
Solubility Information Slightly soluble in water.
Formula Weight 138.14
Percent Purity ≥98%
Chemical Name or Material 3'-Fluoroacetophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.