missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-o-xylene, 99%

Product Code. 11453364
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453364 25 g 25g
11443364 5 g 5g
11463364 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11453364 Supplier Thermo Scientific Alfa Aesar Supplier No. A10637.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 443-82-3 | C8H9F | 124.16 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Stable under normal conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 443-82-3
Molecular Formula C8H9F
Molecular Weight (g/mol) 124.16
MDL Number MFCD00000323
InChI Key AWLDSXJCQWTJPC-UHFFFAOYSA-N
Synonym 3-fluoro-o-xylene, 2,3-dimethylfluorobenzene, 3-fluoro-1,2-xylene, o-xylene, 3-fluoro, 3-fluoro-1,2-dimethylbenzene, benzene, 1-fluoro-2,3-dimethyl, 1,2-dimethyl-3-fluorobenzene, 1-fluoro-2,3-dimethyl-benzene, fluorxylol, pubchem4408
PubChem CID 96489
IUPAC Name 1-fluoro-2,3-dimethylbenzene
SMILES CC1=CC=CC(F)=C1C

Specifications

Density 0.992
Quantity 25 g
Odor Aromatic
Refractive Index 1.486
UN Number UN1993
Beilstein 2040955
Boiling Point 147°C to 148°C
Solubility Information Insoluble in water.
Formula Weight 124.16
Percent Purity 99%
Chemical Name or Material 3-Fluoro-o-xylene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.