missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-N-n-propylbenzylamine, 97%, Thermo Scientific™

Catalog Number 15433338
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
90389-86-9
C10H14FN
167.23
MFCD09938496
OORHPLFSYCEZIP-UHFFFAOYSA-N
n-3-fluorobenzyl propan-1-amine, benzenemethanamine, 3-fluoro-n-propyl, n-3-fluorophenyl methyl propan-1-amine, 3-fluorophenyl methyl propyl amine, 3-fluoro-n-n-propylbenzylamine, 3-fluoro-n-propyl-benzenemethanamine
457580
N-[(3-fluorophenyl)methyl]propan-1-amine
CCCNCC1=CC=CC(F)=C1
This item is not returnable. View return policy
90389-86-9
C10H14FN
167.23
MFCD09938496
OORHPLFSYCEZIP-UHFFFAOYSA-N
n-3-fluorobenzyl propan-1-amine, benzenemethanamine, 3-fluoro-n-propyl, n-3-fluorophenyl methyl propan-1-amine, 3-fluorophenyl methyl propyl amine, 3-fluoro-n-n-propylbenzylamine, 3-fluoro-n-propyl-benzenemethanamine
457580
N-[(3-fluorophenyl)methyl]propan-1-amine
CCCNCC1=CC=CC(F)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3259
Formula Weight 167.22
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Fluoro-N-n-propylbenzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'