missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-DL-valine, 94%, Thermo Scientific Chemicals

Product Code. 11318246
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318246 25 mg 25mg
11328246 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318246 Supplier Thermo Scientific Alfa Aesar Supplier No. L15334.MB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 43163-94-6
Molecular Formula C5H10FNO2
Molecular Weight (g/mol) 135.14
MDL Number MFCD00077549
InChI Key ZFUKCHCGMBNYHH-UHFFFAOYSA-N
Synonym 3-fluoro-dl-valine, 3-fluorovaline, acmc-1cue2
PubChem CID 338887
IUPAC Name 2-amino-3-fluoro-3-methylbutanoic acid
SMILES CC(C)(F)C(N)C(O)=O

Specifications

Melting Point ∽250°C (decomposition)
Beilstein 2350377
Quantity 25 mg
Formula Weight 135.14
Percent Purity 94%
Chemical Name or Material 3-Fluoro-DL-valine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.