missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-(trifluoromethyl)phenylboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 10267647 Shop All Thermo Scientific Chemicals Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
missing translation for 'unitSize'
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10267647 1 g 1g
10411528 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10267647 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 430320010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 159020-59-4
Molecular Formula C7H5BF4O2
Molecular Weight (g/mol) 207.92
MDL Number MFCD06657735
InChI Key WEMCWZGCSRGJGW-UHFFFAOYSA-N
Synonym 3-fluoro-5-trifluoromethyl phenyl boronic acid, 3-fluoro-5-trifluoromethyl phenylboronic acid, 3-fluoro-5-trifluoromethylphenylboronic acid, 3-borono-5-fluorobenzotrifluoride, 3-fluoro-5-trifluoromethyl benzeneboronic acid, 3-fluoro-5-trifluoromethylphenyl boronic acid, 3-trifluoromethyl-5-fluoro-phenylboronic acid, 3-fluoro-5-trifluoromethyl benzene boronic acid, 5-fluoro-3-trifluoromethylbenzeneboronic acid
PubChem CID 2783192
IUPAC Name [3-fluoro-5-(trifluoromethyl)phenyl]boronic acid
SMILES OB(O)C1=CC(F)=CC(=C1)C(F)(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 178.0°C to 180.0°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 207.92
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)phenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • (3-Fluoro-5-trifluoromethylphenyl)boronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.