missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-(trifluoromethyl)phenyl isocyanate, Thermo Scientific™

Product Code. 10331630
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10331630 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10331630 Supplier Thermo Scientific Maybridge Supplier No. MAY00117DA
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 302912-19-2
Packaging Amber Glass Bottle
Molecular Formula C8H3F4NO
Quantity 1 g
Synonym 3-fluoro-5-trifluoromethyl phenyl isocyanate, 1-fluoro-3-isocyanato-5-trifluoromethyl benzene, 3-fluoro-5-trifluoromethyl phenyl, 3-fluoro-5-trifluoromethylphenylisocyanate, 3-fluoro-5-trifluoromethylphenyl isocyanate, 3-fluoro-5-trifluoromethyl phenylisocyanate, 5-fluoro-3-trifluoromethyl benzenisocyanate, 3-fluoro-5-trifluoromethyl-phenylisocyanate, benzene,1-fluoro-3-isocyanato-5-trifluoromethyl
InChI Key WGSXKYXKAARAKD-UHFFFAOYSA-N
SMILES C1=C(C=C(C=C1N=C=O)F)C(F)(F)F
IUPAC Name 1-fluoro-3-isocyanato-5-(trifluoromethyl)benzene
Molecular Weight (g/mol) 205.112
PubChem CID 3445672
Formula Weight 205.11
Percent Purity 97%
Chemical Name or Material 3-fluoro-5-(trifluoromethyl)phenyl isocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.