missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™

Product Code. 11484297
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484297 5 g 5g
11474297 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484297 Supplier Thermo Scientific Alfa Aesar Supplier No. B20385.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 188815-30-7
Molecular Formula C8H4F4O
Molecular Weight (g/mol) 192.113
MDL Number MFCD00061312
InChI Key UZZYXZWSOWQPIS-UHFFFAOYSA-N
Synonym 3-fluoro-5-trifluoromethyl benzaldehyde, 3-fluoro-5-formylbenzotrifluoride, alpha,alpha,alpha,5-tetrafluoro-m-tolualdehyde, benzaldehyde, 3-fluoro-5-trifluoromethyl, pubchem4239, acmc-1bzt1, 5-tetrafluoro-m-tolualdehyde, 3-fluoro-5-trifluromethylbenzaldehyde
PubChem CID 605686
IUPAC Name 3-fluoro-5-(trifluoromethyl)benzaldehyde
SMILES C1=C(C=C(C=C1C(F)(F)F)F)C=O

Specifications

Density 1.376
Boiling Point 175°C
Flash Point 70°C (158°F)
Refractive Index 1.449
Quantity 5 g
Beilstein 9121754
Sensitivity Air Sensitive
Formula Weight 192.11
Percent Purity 97%
Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.