missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-(trifluoromethoxy)benzyl bromide, 97%, Thermo Scientific™

Product Code. p-7032750
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15498885 250 mg 250mg
15437277 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498885 Supplier Thermo Scientific Alfa Aesar Supplier No. H34340.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86256-51-1
Molecular Formula C8H5BrF4O
Molecular Weight (g/mol) 273.025
MDL Number MFCD18783226
InChI Key PJLFSUVIEZRQRU-UHFFFAOYSA-N
Synonym 3-fluoro-5-trifluoromethoxy benzyl bromide, 1-bromomethyl-3-fluoro-5-trifluoromethoxy benzene
PubChem CID 15582616
IUPAC Name 1-(bromomethyl)-3-fluoro-5-(trifluoromethoxy)benzene
SMILES C1=C(C=C(C=C1OC(F)(F)F)F)CBr

Specifications

Quantity 250 mg
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 273.02
Percent Purity 97%
Chemical Name or Material 3-Fluoro-5-(trifluoromethoxy)benzyl bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.