missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-(pentafluorothio)aniline, 97%

Product Code. 15466537
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15466537 250 mg 250mg
15456537 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15466537 Supplier Thermo Scientific Alfa Aesar Supplier No. H33680.MD
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1240256-99-8 | C6H5F6NS | 237.163 g/mol

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1240256-99-8
Molecular Formula C6H5F6NS
Molecular Weight (g/mol) 237.163
MDL Number MFCD16652430
InChI Key RFXWDLYDFAZNLO-UHFFFAOYSA-N
Synonym 3-fluoro-5-pentafluorosulfur aniline, 3-fluoro-5-pentafluoro-??-sulfanyl aniline, 3-fluoro-5-pentafluorothio aniline
PubChem CID 66523473
IUPAC Name 3-fluoro-5-(pentafluoro-$l^{6}-sulfanyl)aniline
SMILES C1=C(C=C(C=C1F)S(F)(F)(F)(F)F)N

Specifications

Quantity 250 mg
UN Number UN2811
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 237.16
Percent Purity 97%
Chemical Name or Material 3-Fluoro-5-(pentafluorothio)aniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.