missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid, 95%, Thermo Scientific Chemicals

Catalog Number 15400366
Click to view available options
Quantity:
250 mg
This item is not returnable. View return policy
871329-62-3
C8H8BFO4
197.956
MFCD07363749
VKHJVASTGLPBBL-UHFFFAOYSA-N
3-fluoro-5-methoxycarbonyl phenyl boronic acid, 3-fluoro-5-methoxycarbonylphenyl boronic acid, 3-fluoro-5-methoxycarbonyl phenylboronic acid, 3-fluoro-5-methoxycarbonyl benzeneboronic acid, benzoic acid,3-borono-5-fluoro-, 1-methyl ester 9ci, methyl 3-borono-5-fluorobenzoate, acmc-209qg6, ksc657s1f, 3-fluoro-5-methoxycarbonylphenylboronicacid
44717206
(3-fluoro-5-methoxycarbonylphenyl)boronic acid
B(C1=CC(=CC(=C1)F)C(=O)OC)(O)O
This item is not returnable. View return policy
871329-62-3
C8H8BFO4
197.956
MFCD07363749
VKHJVASTGLPBBL-UHFFFAOYSA-N
3-fluoro-5-methoxycarbonyl phenyl boronic acid, 3-fluoro-5-methoxycarbonylphenyl boronic acid, 3-fluoro-5-methoxycarbonyl phenylboronic acid, 3-fluoro-5-methoxycarbonyl benzeneboronic acid, benzoic acid,3-borono-5-fluoro-, 1-methyl ester 9ci, methyl 3-borono-5-fluorobenzoate, acmc-209qg6, ksc657s1f, 3-fluoro-5-methoxycarbonylphenylboronicacid
44717206
(3-fluoro-5-methoxycarbonylphenyl)boronic acid
B(C1=CC(=CC(=C1)F)C(=O)OC)(O)O

3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 138°C to 142°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 197.96
Percent Purity 95%
Chemical Name or Material 3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'