missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-hydroxypyridine, 97%, Thermo Scientific™

Catalog Number 15495427
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
209328-55-2
C5H4FNO
113.09
MFCD03095199
IUFLFKASIHPKNZ-UHFFFAOYSA-N
3-fluoro-5-hydroxypyridine, 5-fluoro-3-pyridinol, 3-pyridinol, 5-fluoro, 5-fluoro-3-hydroxypyridine, pubchem6625, acmc-209ffy, 5-fluoranylpyridin-3-ol, ksc201g7t, 3-pyridinol, 5-fluoro-9ci
2762906
5-fluoropyridin-3-ol
OC1=CC(F)=CN=C1
This item is not returnable. View return policy
209328-55-2
C5H4FNO
113.09
MFCD03095199
IUFLFKASIHPKNZ-UHFFFAOYSA-N
3-fluoro-5-hydroxypyridine, 5-fluoro-3-pyridinol, 3-pyridinol, 5-fluoro, 5-fluoro-3-hydroxypyridine, pubchem6625, acmc-209ffy, 5-fluoranylpyridin-3-ol, ksc201g7t, 3-pyridinol, 5-fluoro-9ci
2762906
5-fluoropyridin-3-ol
OC1=CC(F)=CN=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 161°C to 164°C
Quantity 250 mg
Formula Weight 113.09
Percent Purity 97%
Chemical Name or Material 3-Fluoro-5-hydroxypyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'