missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific™

Product Code. 11414427
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11414427

Brand: Thermo Scientific Alfa Aesar B20454.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as an active pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Moisture sensitive. Store away from alcohols, amines, oxidizing agents and active metals. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 216144-68-2
Molecular Formula C8H3ClF4O
Molecular Weight (g/mol) 226.55
MDL Number MFCD00236284
InChI Key DPDAEMNQJYDHRQ-UHFFFAOYSA-N
Synonym 3-fluoro-4-trifluoromethyl benzoyl chloride, 3-fluoro-4-trifluoromethyl benzoylchloride, 3-fluoranyl-4-trifluoromethyl benzoyl chloride, acmc-1cobr, 4-chlorocarbonyl-2-fluorobenzotrifluoride, 3-fluoro-4-trifiuoromethyl benzoyl chloride, 3-fluoro-4-trifluoromethyl benzoyl cloride, benzoylchloride, 3-fluoro-4-trifluoromethyl, benzoyl chloride, 3-fluoro-4-trifluoromethyl
PubChem CID 2734709
IUPAC Name 3-fluoro-4-(trifluoromethyl)benzoyl chloride
SMILES FC1=CC(=CC=C1C(F)(F)F)C(Cl)=O

Specifications

Density 1.486
Boiling Point 151°C to 153°C
Flash Point 81°C (177°F)
Refractive Index 1.4656
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 226.56
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-(trifluoromethyl)benzoyl chloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.