missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-(trifluoromethyl)benzoic acid, TRC

Product Code. 30390152
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30390152 250 mg
30405760 500 mg
30405947 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390152 Supplier Toronto Research Chemicals Supplier No. TRCF598468250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115754-21-7
Molecular Formula C8H4F4O2
Molecular Weight (g/mol) 208.11
SMILES OC(=O)c1ccc(c(F)c1)C(F)(F)F

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C8H4F4O2/c9-6-3-4(7(13)14)1-2-5(6)8(10,11)12/h1-3H,(H,13,14)
Formula Weight 208.0147
Chemical Name or Material 3-Fluoro-4-(trifluoromethyl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.