missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-(trifluoromethyl)benzoic acid, 97+%, Thermo Scientific™

Product Code. 11490488
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490488 5 g 5g
11480488 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490488 Supplier Thermo Scientific Alfa Aesar Supplier No. B24625.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115754-21-7
Molecular Formula C8H4F4O2
Molecular Weight (g/mol) 208.112
MDL Number MFCD00236279
InChI Key HRIHSNPFVGMAKX-UHFFFAOYSA-N
Synonym 3-fluoro-4-trifluoromethyl benzoic acid, benzoic acid, 3-fluoro-4-trifluoromethyl, alpha,alpha,alpha,3-tetrafluoro-p-toluic acid, pubchem4970, acmc-1c70e, ksc179k4t, 3-fluoro-4 trifluoromethyl benzoic acid, 3-fluoro-4-trifluoromethyl-benzoic acid, 3-fluoro-4-trifluoromethyl benzoicacid, 3-fluoro-4-trifluoromethyl benzoic aicd
PubChem CID 688293
IUPAC Name 3-fluoro-4-(trifluoromethyl)benzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)F)C(F)(F)F

Specifications

Melting Point 173°C to 176°C
Quantity 5 g
Formula Weight 208.11
Percent Purity ≥97%
Chemical Name or Material 3-Fluoro-4-(trifluoromethyl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.