missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-(trifluoromethyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Product Code. 15487866
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15487866 1 g 1g
15497866 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15487866 Supplier Thermo Scientific Alfa Aesar Supplier No. H62811.03
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 445303-67-3 | Molecular Formula: C13H15BF4O2 | Molecular Weight: 290.065 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 445303-67-3
Molecular Formula C13H15BF4O2
Molecular Weight (g/mol) 290.065
MDL Number MFCD09878544
InChI Key FPCSARGPMNDYAO-UHFFFAOYSA-N
Synonym 2-3-fluoro-4-trifluoromethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-fluoro-4-trifluoromethyl phenylboronic acid pinacol ester, 3-fluoro-4-trifluoromethyl phenylboronic acid, pinacol ester, 3-fluoro-4-trifluoromethyl benzeneboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-3-fluoro-4-trifluoromethyl phenyl-4,4,5,5-tetramethyl, 2-3-fluoro-4-trifluoromethyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxa-borolane
PubChem CID 46739054
IUPAC Name 2-[3-fluoro-4-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)C(F)(F)F)F

Specifications

Quantity 1 g
Formula Weight 290.06
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Fluoro-4-(trifluoromethyl)benzeneboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.