missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-nitrophenol, 99%

Product Code. 10217610 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10217610 5 g Glass bottle 5g
10662145 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10217610 Supplier Thermo Scientific Acros Supplier No. 303370050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 394-41-2 | C6H4FNO3 | 157.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 394-41-2
Molecular Formula C6H4FNO3
Molecular Weight (g/mol) 157.1
MDL Number MFCD00041251
InChI Key CSSGKHVRDGATJL-UHFFFAOYSA-N
Synonym phenol, 3-fluoro-4-nitro, 3-fluoro-4-nitro-phenol, 2-fluoro-4-hydroxynitrobenzene, pubchem2101, 3-fluoro4-nitrophenol, 4-nitro-3-fluorophenol, 5-fluoro-4-nitrophenol, acmc-1afpw, 3-fluoro-4-nitro phenol, ksc490k8p
PubChem CID 520948
IUPAC Name 3-fluoro-4-nitrophenol
SMILES C1=CC(=C(C=C1O)F)[N+](=O)[O-]

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 92°C to 95°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H3(NO2)OH
Quantity 5 g
Formula Weight 157.1
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3-Fluoro-4-nitrophenol, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.