missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-nitrobenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog Number p-7028785
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
131858-37-2
C7H5BrFNO2
234.02
MFCD03094237
ZOZJSWIXPIVMRU-UHFFFAOYSA-N
3-fluoro-4-nitrobenzyl bromide, 4-bromomethyl-2-fluoro-1-nitrobenzene, 4-bromomethyl-2-fluoronitrobenzene, benzene, 4-bromomethyl-2-fluoro-1-nitro, intermediates-zcf02012, acmc-1c6h7, 4-bromomethyl-2-fluoro-1-nitro-benzene, benzene,4-bromomethyl-2-fluoro-1-nitro, 4-bromomethyl-2-fluoranyl-1-nitro-benzene
2774661
4-(bromomethyl)-2-fluoro-1-nitrobenzene
[O-][N+](=O)C1=C(F)C=C(CBr)C=C1
This item is not returnable. View return policy
131858-37-2
C7H5BrFNO2
234.02
MFCD03094237
ZOZJSWIXPIVMRU-UHFFFAOYSA-N
3-fluoro-4-nitrobenzyl bromide, 4-bromomethyl-2-fluoro-1-nitrobenzene, 4-bromomethyl-2-fluoronitrobenzene, benzene, 4-bromomethyl-2-fluoro-1-nitro, intermediates-zcf02012, acmc-1c6h7, 4-bromomethyl-2-fluoro-1-nitro-benzene, benzene,4-bromomethyl-2-fluoro-1-nitro, 4-bromomethyl-2-fluoranyl-1-nitro-benzene
2774661
4-(bromomethyl)-2-fluoro-1-nitrobenzene
[O-][N+](=O)C1=C(F)C=C(CBr)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 46°C to 48°C
Quantity 1 g
UN Number UN3261
Formula Weight 234.02
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-nitrobenzyl bromide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'