missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluoro-4-methylphenylmagnesium bromide, 0.50 M in 2-MeTHF, Thermo Scientific™
Click to view available options
Quantity:
100 mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Solubility
Reacts with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. It is sensitive to air and moisture. Protect from humidity
Reacts with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. It is sensitive to air and moisture. Protect from humidity
Specifications
CAS | 185077-02-5 |
Molecular Formula | C7H6BrFMg |
MDL Number | MFCD01311470 |
Quantity | 100 mL |
UN Number | UN2924 |
Sensitivity | Air and moisture sensitive |
Synonym | 3-fluoro-4-methylphenylmagnesium bromide, bromo 3-fluoro-4-methylphenyl magnesium, 3-fluoro-4-methylphenylmagnesium bromide 0.5m solution in thf, fgtairviejuzjo-uhfffaoysa-m, 3-fluoro-4-methylphenyl magnesiumbromide, magnesium,bromo 3-fluoro-4-methylphenyl, 3-fluoro-4-methylphenylmagnesium bromide, 0.50 m in 2-methf, 3-fluoro-4-methylphenylmagnesium bromide 0.5 m in tetrahydrofuran, 3-fluoro-4-methylphenylmagnesium bromide solution, 0.5 m in thf, grignard reagent |
Solubility Information | Reacts with water. |
InChI Key | FGTAIRVIEJUZJO-UHFFFAOYSA-M |
SMILES | CC1=CC=C([Mg]Br)C=C1F |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'