missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methylphenylboronic acid, 97%, Thermo Scientific™

Product Code. 10785262 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10785262 1 g Glass bottle 1g
10179359 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10785262 Supplier Thermo Scientific Acros Supplier No. 433420010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 168267-99-0
Molecular Formula C7H8BFO2
Molecular Weight (g/mol) 153.95
MDL Number MFCD02683115
InChI Key WPVBHUUZDFUIJA-UHFFFAOYSA-N
Synonym 3-fluoro-4-methylbenzeneboronic acid, 3-fluoro-4-methylphenyl boronic acid, 3-fluoro-p-tolylboronic acid, 3-fluoro-4-methylphenyl-boronic acid, boronic acid, 3-fluoro-4-methylphenyl, pubchem5124, 3-fluoro-4-methyl-phenyl boronic acid, acmc-209dxx, ksc173s0n
PubChem CID 2782674
IUPAC Name (3-fluoro-4-methylphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)C)F)(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 241.0°C to 244.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 153.95
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-4-methylphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Fluoro-4-methylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.