missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methylbenzyl alcohol, 97%, Thermo Scientific™

Product Code. 11344056
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11344056 250 mg 250mg
11334056 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11344056 Supplier Thermo Scientific Alfa Aesar Supplier No. L19170.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 192702-79-7
Molecular Formula C8H9FO
Molecular Weight (g/mol) 140.16
MDL Number MFCD01631533
InChI Key UUVJAGTYIVYFOS-UHFFFAOYSA-N
PubChem CID 2774590
IUPAC Name (3-fluoro-4-methylphenyl)methanol
SMILES CC1=CC=C(CO)C=C1F

Specifications

Melting Point 33°C to 36°C
Quantity 250 mg
Beilstein 7700178
Synonym 3-fluoro-4-methylphenyl methanol, 3-fluoro-4-methylbenzyl alcohol, benzenemethanol, 3-fluoro-4-methyl, benzenemethanol,3-fluoro-4-methyl, benzenemethanol, 3-fluoro-4-methyl-9ci, 3-fluoro-4-methylphenyl methan-1-ol, acmc-20an3o, rarechem al bd 0500, 3-fluoro-4-methyl-phenyl-methanol, 3-fluoranyl-4-methyl-phenyl methanol
Formula Weight 140.16
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-methylbenzyl alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.