missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methylbenzonitrile, 98%, Thermo Scientific™

Product Code. 11435646
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 11435646

Brand: Thermo Scientific Alfa Aesar B20764.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 170572-49-3
Melting Point 45°C to 49°C
Boiling Point 204°C
Flash Point 83°C (181°F)
Molecular Formula C8H6FN
MDL Number MFCD00153174
Quantity 10 g
UN Number UN3439
Beilstein 7700177
Synonym 4-cyano-2-fluorotoluene, 3-fluoro-p-tolunitrile, 2-fluoro-4-cyanotoluene, benzonitrile, 3-fluoro-4-methyl, 4-methyl-3-fluorobenzonitrile, 3-fluoro-4-methyl-benzonitrile, 3-fluoro-4-methylbenzenecarbonitrile, benzonitrile, 3-fluoro-4-methyl-9ci, ncr cf d1, pubchem1557
InChI Key KUQQONVKIURIQU-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1F)C#N
IUPAC Name 3-fluoro-4-methylbenzonitrile
Molecular Weight (g/mol) 135.14
PubChem CID 585140
Formula Weight 135.14
Percent Purity 98%
Chemical Name or Material 3-Fluoro-4-methylbenzonitrile
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.