missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methyl(thiobenzamide), 97%, Thermo Scientific™

Catalog Number 15423477
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
175277-87-9
C8H8FNS
169.217
MFCD00275549
WFCXNRMNVWYANI-UHFFFAOYSA-N
3-fluoro-4-methylbenzene-1-carbothioamide, 3-fluoro-4-methylthiobenzamide, 3-fluoro-4-methyl-thiobenzamide, 3-fluoro-4-methylbenzothioamide, benzenecarbothioamide,3-fluoro-4-methyl, 3-fluoranyl-4-methyl-benzenecarbothioamide, amino 3-fluoro-4-methylphenyl methane-1-thione, benzenecarbothioamide, 3-fluoro-4-methyl-9ci
3823232
3-fluoro-4-methylbenzenecarbothioamide
CC1=C(C=C(C=C1)C(=S)N)F
This item is not returnable. View return policy
175277-87-9
C8H8FNS
169.217
MFCD00275549
WFCXNRMNVWYANI-UHFFFAOYSA-N
3-fluoro-4-methylbenzene-1-carbothioamide, 3-fluoro-4-methylthiobenzamide, 3-fluoro-4-methyl-thiobenzamide, 3-fluoro-4-methylbenzothioamide, benzenecarbothioamide,3-fluoro-4-methyl, 3-fluoranyl-4-methyl-benzenecarbothioamide, amino 3-fluoro-4-methylphenyl methane-1-thione, benzenecarbothioamide, 3-fluoro-4-methyl-9ci
3823232
3-fluoro-4-methylbenzenecarbothioamide
CC1=C(C=C(C=C1)C(=S)N)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 177°C to 179°C (decomposition)
Quantity 250 mg
UN Number UN2811
Formula Weight 169.22
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-methyl(thiobenzamide)
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'