missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methoxypyridine, 97%

Product Code. 15461879
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15461879 1 g 1g
15481879 250 mg 250mg
15471879 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15461879 Supplier Thermo Scientific Alfa Aesar Supplier No. H64603.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1060805-03-9 | C6H6FNO | 127.118 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1060805-03-9
Molecular Formula C6H6FNO
Molecular Weight (g/mol) 127.118
MDL Number MFCD13185605
InChI Key XYTUUTJNWFECNS-UHFFFAOYSA-N
Synonym 3-fluoro-4-methoxy-pyridine, pyridine, 3-fluoro-4-methoxy, abbypharma ap-14-5665
PubChem CID 57336639
IUPAC Name 3-fluoro-4-methoxypyridine
SMILES COC1=C(C=NC=C1)F

Specifications

Quantity 1 g
Beilstein 19561531
Formula Weight 127.12
Percent Purity 98%
Chemical Name or Material 3-Fluoro-4-methoxypyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.