missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-(methoxycarbonyl)benzeneboronic acid, 97%

Product Code. p-7034811
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15429865

Brand: Thermo Scientific Alfa Aesar H52510.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 505083-04-5 | C8H8BFO4 | 197.956 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 505083-04-5
Molecular Formula C8H8BFO4
Molecular Weight (g/mol) 197.956
MDL Number MFCD04115665
InChI Key YZYGXFXSMDUXJT-UHFFFAOYSA-N
Synonym 3-fluoro-4-methoxycarbonyl phenyl boronic acid, 3-fluoro-4-methoxycarbonyl phenylboronic acid, 3-fluoro-4-methoxycarbonyl benzeneboronic acid, methyl 4-borono-2-fluorobenzoate, 4-methoxycarbonyl-3-fluorophenylboronic acid, pubchem17771, 3-fluoro-4-methoxycarboxyphenylboronicacid, acmc-1atlt, 3-fluoro-4-methoxycarbonyl-phenyl boronic acid
PubChem CID 2782818
IUPAC Name (3-fluoro-4-methoxycarbonylphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)C(=O)OC)F)(O)O

Specifications

Melting Point 180°C to 184°C
Quantity 250 mg
Formula Weight 197.96
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-(methoxycarbonyl)benzeneboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.