missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methoxybenzyl bromide, 98%

Product Code. p-7028711
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15403397

Brand: Thermo Scientific Alfa Aesar H26116.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 331-61-3 | C8H8BrFO | 219.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 331-61-3
Molecular Formula C8H8BrFO
Molecular Weight (g/mol) 219.05
MDL Number MFCD00671768
InChI Key KLWYYWVSFYRPLM-UHFFFAOYSA-N
Synonym 3-fluoro-4-methoxybenzyl bromide, 4-bromomethyl-2-fluoro-1-methoxybenzene, 3-fluoro-4-methoxybenzylbromide, 4-bromomethyl-2-fluoro-1-methoxy-benzene, benzene, 4-bromomethyl-2-fluoro-1-methoxy, pubchem4923, acmc-1ckmo, 4-bromomethyl-2-fluoroanisole
PubChem CID 2774553
IUPAC Name 4-(bromomethyl)-2-fluoro-1-methoxybenzene
SMILES COC1=CC=C(CBr)C=C1F

Specifications

Melting Point 44°C to 46°C
Odor Pungent
Quantity 250 mg
UN Number UN3261
Formula Weight 219.05
Percent Purity 98%
Chemical Name or Material 3-Fluoro-4-methoxybenzyl bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.