missing translation for 'onlineSavingsMsg'
Learn More

3'-Fluoro-4'-methoxyacetophenone, 99%

Product Code. 11463208
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463208 25 g 25g
11453208 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11463208 Supplier Thermo Scientific Alfa Aesar Supplier No. A18972.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 455-91-4 | C9H9FO2 | 168.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 455-91-4
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.167
MDL Number MFCD00026219
InChI Key LQASUDVYVOFKNK-UHFFFAOYSA-N
Synonym 3'-fluoro-4'-methoxyacetophenone, 3-fluoro-4-methoxyacetophenone, 1-3-fluoro-4-methoxyphenyl ethanone, 1-3-fluoro-4-methoxyphenyl ethan-1-one, ethanone, 1-3-fluoro-4-methoxyphenyl, 3'-fluoro-4-methoxyacetophenone, acetophenone, 3'-fluoro-4'-methoxy, 1-3-fluoro-4-methoxy-phenyl-ethanone, 1-3-fluoro-4-methoxyphenyl-1-ethanone, 1-3-fluoro-4-methoxy-phenyl ethanone
PubChem CID 96828
IUPAC Name 1-(3-fluoro-4-methoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)OC)F

Specifications

Melting Point 90°C to 94°C
Boiling Point 147°C to 148°C (20 mmHg)
Quantity 25 g
Beilstein 2084062
Formula Weight 168.17
Percent Purity 99%
Chemical Name or Material 3'-Fluoro-4'-methoxyacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.