missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-hydroxyphenylacetic acid, 98%

Product Code. 11400053
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400053 5 g 5g
11490043 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11400053 Supplier Thermo Scientific Alfa Aesar Supplier No. A12114.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 458-09-3 | C8H7FO3 | 170.139 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 458-09-3
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.139
MDL Number MFCD00004348
InChI Key YRFBZAHYMOSSGX-UHFFFAOYSA-N
Synonym 3-fluoro-4-hydroxyphenylacetic acid, 2-3-fluoro-4-hydroxyphenyl acetic acid, 3-fluoro-4-hydroxyphenyl acetic acid, 3-fluoro-4-hydroxybenzeneacetic acid, benzeneacetic acid, 3-fluoro-4-hydroxy, 2-3-fluoro-4-hydroxy-phenyl acetic acid, 4-carboxymethyl-2-fluorophenol, 3-fluoro-4-hydroxy-phenyl-acetic acid, acmc-1ahjh, opera_id_1839
PubChem CID 68014
IUPAC Name 2-(3-fluoro-4-hydroxyphenyl)acetic acid
SMILES C1=CC(=C(C=C1CC(=O)O)F)O

Specifications

Melting Point 130°C to 133°C
Quantity 5 g
Formula Weight 170.14
Percent Purity 98%
Chemical Name or Material 3-Fluoro-4-hydroxyphenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.