missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-hydroxy-5-methoxybenzaldehyde, TRC

Product Code. 30392971
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392971 250 mg
30397014 1 g
30400365 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392971 Supplier Toronto Research Chemicals Supplier No. TRCF591750250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79418-78-3
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.14
Synonym 3-Fluoro-4-hydroxy-5-methoxybenzaldehyde, 3-Fluoro-5-methoxy-4-hydroxybenzaldehyde, 5-Fluoro-4-hydroxy-3-methoxybenzaldehyde, 5-Fluorovanillin
SMILES COc1cc(C=O)cc(F)c1O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C8H7FO3/c1-12-7-3-5(4-10)2-6(9)8(7)11/h2-4,11H,1H3
Formula Weight 170.0379
Chemical Name or Material 3-Fluoro-4-hydroxy-5-methoxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.