missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-(4-methyl-1-piperazinyl)benzeneboronic acid pinacol ester, 95%

Product Code. 15495146
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15495146 1 g 1g
15405156 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15495146 Supplier Thermo Scientific Alfa Aesar Supplier No. H59640.03
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1408088-34-5 | C17H26BFN2O2 | 320.215 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1408088-34-5
Molecular Formula C17H26BFN2O2
Molecular Weight (g/mol) 320.215
MDL Number MFCD22988988
InChI Key RFECQDXNBWGCRR-UHFFFAOYSA-N
Synonym 1-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-4-methylpiperazine, 3-fluoro-4-4-methyl-1-piperazinyl benzeneboronic acid pinacol ester
PubChem CID 70974710
IUPAC Name 1-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-4-methylpiperazine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)N3CCN(CC3)C)F

Specifications

Quantity 1 g
Formula Weight 320.21
Percent Purity 95%
Chemical Name or Material 3-Fluoro-4-(4-methyl-1-piperazinyl)benzeneboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.