missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-nitropyridine, 96%

Product Code. 15406687
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15406687 1 g 1g
15426687 250 mg 250mg
15416687 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15406687 Supplier Thermo Scientific Alfa Aesar Supplier No. H33813.03
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 54231-35-5 | C5H3FN2O2 | 142.089 g/mol

A specialty product for proteomics research applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A specialty product for proteomics research applications.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54231-35-5
Molecular Formula C5H3FN2O2
Molecular Weight (g/mol) 142.089
MDL Number MFCD04114127
InChI Key IJVFHCSUEBAAOZ-UHFFFAOYSA-N
Synonym 2-nitro-3-fluoropyridine, 3-fluoro-2-nitro-pyridine, pyridine, 3-fluoro-2-nitro, pubchem2185, 3-fluor-2-nitropyridin, pyfn51, 3-fluoro 2-nitro pyridine, pyridine,3-fluoro-2-nitro, pyridine, 3-fluoro-2-nitro-9ci
PubChem CID 2762802
IUPAC Name 3-fluoro-2-nitropyridine
SMILES C1=CC(=C(N=C1)[N+](=O)[O-])F

Specifications

Melting Point 30°C to 35°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 142.09
Percent Purity 96%
Chemical Name or Material 3-Fluoro-2-nitropyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.