missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluoro-2-methylpyridine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
3-Fluoro-2-methylpyridine was used in the synthesis of fluorine-containing pyridinealdoximes of potential use for the reactivation of cholinesterase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Fluoro-2-methylpyridine was used in the synthesis of fluorine-containing pyridinealdoximes of potential use for the reactivation of cholinesterase.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
3-Fluoro-2-methylpyridine was used in the synthesis of fluorine-containing pyridinealdoximes of potential use for the reactivation of cholinesterase.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Color | Colorless to Yellow |
Quantity | 250 mg |
Beilstein | 6497832 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 111.12 |
Percent Purity | 98% |
Physical Form | Liquid |
Chemical Name or Material | 3-Fluoro-2-methylpyridine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'