missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-methoxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number 15437706
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1417301-67-7
C13H18BFO3
252.092
OONNDCZIVBSYTG-UHFFFAOYSA-N
2-3-fluoro-2-methoxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-fluoro-2-methoxybenzeneboronic acid pinacol ester, 3-fluoro-2-methoxyphenylboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-3-fluoro-2-methoxyphenyl-4,4,5,5-tetramethyl
72221148
2-(3-fluoro-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)F)OC
This item is not returnable. View return policy
1417301-67-7
C13H18BFO3
252.092
OONNDCZIVBSYTG-UHFFFAOYSA-N
2-3-fluoro-2-methoxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-fluoro-2-methoxybenzeneboronic acid pinacol ester, 3-fluoro-2-methoxyphenylboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-3-fluoro-2-methoxyphenyl-4,4,5,5-tetramethyl
72221148
2-(3-fluoro-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)F)OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 252.09
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Fluoro-2-methoxybenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'