missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-methoxyaniline, 97%, Thermo Scientific Chemicals

Product Code. p-7031217
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15444907

Brand: Thermo Scientific Alfa Aesar H32214.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 437-83-2
Molecular Formula C7H8FNO
Molecular Weight (g/mol) 141.145
MDL Number MFCD00007759
InChI Key RCYMPYMITUEHOJ-UHFFFAOYSA-N
Synonym 3-fluoro-o-anisidine, 3-fluoro-2-methoxy-phenylamine, 2-amino-6-fluoroanisole, 3-fluoro-2-methoxyphenylamine, benzenamine, 3-fluoro-2-methoxy, 3-fuloro-2-methoxyaniline, 3-fluoro-2-methoxy-benzenamine, 3-fluoro-2-methoxybenzenenamine, pubchem2256, pubchem3021
PubChem CID 581109
IUPAC Name 3-fluoro-2-methoxyaniline
SMILES COC1=C(C=CC=C1F)N

Specifications

Quantity 250 mg
UN Number UN2810
Formula Weight 141.15
Percent Purity 97%
Chemical Name or Material 3-Fluoro-2-methoxyaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.