missing translation for 'onlineSavingsMsg'
Learn More

3-exo-Aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxamide, 99+%, Thermo Scientific Chemicals

Product Code. 10308763 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10308763 1 g Glass Bottle 1g
10627441 250 mg Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10308763 Supplier Thermo Scientific Acros Supplier No. 277810010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 105786-40-1
Molecular Formula C8H13N2O
Molecular Weight (g/mol) 153.20
MDL Number MFCD00143157,MFCD01456476,MFCD00143157
InChI Key SCQSHSJVMGGQKR-UCROKIRRSA-O
Synonym 3-exo-aminobicyclo 2.2.1 hept-5-ene-2-carboxamide, +-1s,2s,3r,4r-3-aminobicyclo 2.2.1 hept-5-ene-2-carboxamide, +/--1s,2s,3r,4r-3-aminobicyclo 2.2.1 hept-5-ene-2-carboxamide, bicyclo 2.2.1 hept-5-ene-2-carboxamide, 3-amino-, 1r,2r,3s,4s-rel-9ci
PubChem CID 42580098
IUPAC Name (1S,2S,3R,4R)-3-aminobicyclo[2.2.1]hept-5-ene-2-carboxamide
SMILES NC(=O)[C@H]1[C@@H]([NH3+])[C@@H]2C[C@H]1C=C2

Spécification

Melting Point 235.0°C to 253.0°C
CAS Min % 99.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Solubility Information Solubility in water: .. Other solubilities: soluble in ethanol and methanol
Quantity 1 g
Formula Weight 152.2
Percent Purity ≥99%
Physical Form Crystalline Powder
Chemical Name or Material 3-exo-Aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxamide, 99+%
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.