missing translation for 'onlineSavingsMsg'
Learn More

3-Ethynylbenzeneboronic acid pinacol ester, 95%, Thermo Scientific™

Product Code. 15498387
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15498387 1 g 1g
15408397 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498387 Supplier Thermo Scientific Alfa Aesar Supplier No. H51902.03
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 946168-04-3
Molecular Formula C14H17BO2
Molecular Weight (g/mol) 228.098
MDL Number MFCD16294529
InChI Key SJDFLZSEQGKSDJ-UHFFFAOYSA-N
Synonym 2-3-ethynylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-3-ethynyl-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 3-ethynylbenzeneboronic acid pinacol ester, amtb210, 3-ethynylphenyl boronic acid pinacol ester, 2-3-ethynyl-phenyl-4,4,5,5-tetramethyl-1,3,2dioxaborolane
PubChem CID 57416900
IUPAC Name 2-(3-ethynylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C#C

Specifications

Quantity 1 g
Beilstein 11140327
Formula Weight 228.1
Percent Purity 95%
Chemical Name or Material 3-Ethynylbenzeneboronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.