missing translation for 'onlineSavingsMsg'
Learn More

3-Ethynylaniline, TRC

Product Code. 30134673
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134673 1 g 1g
30133678 10 mg 10mg
17953175 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134673 Supplier Toronto Research Chemicals Supplier No. TRCE6850701G
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54060-30-9
Molecular Formula C8H7N
Molecular Weight (g/mol) 117.15
Synonym (3-Ethynylphenyl)amine, (m-Aminophenyl)acetylene, 1-Amino-3-ethynylbenzene, 3-Acetylenylaniline-d4, 3-Amino-1-ethynylbenzene, 3-Ethynylaniline, 3-Ethynylbenzenamine, m-Ethynylaniline;
SMILES C#CC1=CC=CC(=C1)N

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H7N/c1-2-7-4-3-5-8(9)6-7/h1,3-6H,9H2
Chemical Name or Material 3-Ethynylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.