missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyltoluene, 97%

Product Code. 11399515
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399515 25 g 25g
11389515 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399515 Supplier Thermo Scientific Alfa Aesar Supplier No. L18788.14
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 620-14-4 | C9H12 | 120.20 g/mol

It is used in the formation of 3-ethylsalicylic acid from 3-Ethyltoluene by Pseudomonas ovalis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the formation of 3-ethylsalicylic acid from 3-Ethyltoluene by Pseudomonas ovalis.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away. Protect from electrostatic charges.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-14-4
Molecular Formula C9H12
Molecular Weight (g/mol) 120.20
MDL Number MFCD00009259
InChI Key ZLCSFXXPPANWQY-UHFFFAOYSA-N
Synonym 3-ethyltoluene, m-ethyltoluene, 1-methyl-3-ethylbenzene, benzene, 1-ethyl-3-methyl, m-ethylmethylbenzene, m-methylethylbenzene, toluene, m-ethyl, 3-methylethylbenzene, 1-ethyl-3-methyl-benzene, unii-737ptd7o7e
PubChem CID 12100
ChEBI CHEBI:77512
IUPAC Name 1-ethyl-3-methylbenzene
SMILES CCC1=CC=CC(C)=C1

Specifications

Melting Point -96°C
Density 0.865
Boiling Point 158°C to 159°C
Flash Point 38°C (100°F)
Refractive Index 1.496
Quantity 25 g
UN Number UN3295
Beilstein 1850821
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 120.2
Percent Purity ≥98%
Chemical Name or Material 3-Ethyltoluene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.