missing translation for 'onlineSavingsMsg'
Learn More

3-Ethylphenol, 95%

Product Code. 11446420
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11446420 50 g 50g
11436420 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11446420 Supplier Thermo Scientific Alfa Aesar Supplier No. B23470.18

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 620-17-7 | C8H10O | 122.17 g/mol

It is used as a photographic chemical intermediate and an intermediate for the cyan coupler of photographic paper. It has a wide application in fragrance industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a photographic chemical intermediate and an intermediate for the cyan coupler of photographic paper. It has a wide application in fragrance industry.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-17-7
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002311
InChI Key HMNKTRSOROOSPP-UHFFFAOYSA-N
Synonym m-ethylphenol, phenol, 3-ethyl, phenol, m-ethyl, meta-ethylphenol, 1-ethyl-3-hydroxybenzene, 1-hydroxy-3-ethylbenzene, 3-ethyl-phenol, unii-0g9zk222jx, benzene, 1-ethyl-3-hydroxy, dsstox_cid_2480
PubChem CID 12101
ChEBI CHEBI:34332
IUPAC Name 3-ethylphenol
SMILES CCC1=CC=CC(O)=C1

Specifications

Melting Point -4°C
Density 1.028
Boiling Point 218°C
Flash Point 94°C (202°F)
Refractive Index 1.5335
Quantity 50 g
UN Number UN3145
Beilstein 1857002
Solubility Information Slightly soluble in water.
Formula Weight 122.17
Percent Purity 95%
Chemical Name or Material 3-Ethylphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.