missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Ethylphenol, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
It is used as a photographic chemical intermediate and an intermediate for the cyan coupler of photographic paper. It has a wide application in fragrance industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a photographic chemical intermediate and an intermediate for the cyan coupler of photographic paper. It has a wide application in fragrance industry.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
It is used as a photographic chemical intermediate and an intermediate for the cyan coupler of photographic paper. It has a wide application in fragrance industry.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Specifications
Melting Point | -4°C |
Density | 1.028 |
Boiling Point | 218°C |
Flash Point | 94°C (202°F) |
Refractive Index | 1.5335 |
Quantity | 10 g |
UN Number | UN3145 |
Beilstein | 1857002 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 122.17 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'