missing translation for 'onlineSavingsMsg'
Learn More

3-Ethylaniline, 97%

Product Code. 10360954 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10360954 250 g Glass bottle 250g
10753561 10 g Glass bottle 10g
10760681 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10360954 Supplier Thermo Scientific Acros Supplier No. 174552500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 587-02-0 | C8H11N | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 587-02-0
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00007818
InChI Key AMKPQMFZCBTTAT-UHFFFAOYSA-N
Synonym m-ethylaniline, benzenamine, 3-ethyl, 3-ethylbenzenamine, 3-ethylphenylamine, aniline, m-ethyl, unii-w5u3oqy77o, aniline, 3-ethyl, w5u3oqy77o, methylanilin, 3-ethyl aniline
PubChem CID 11475
IUPAC Name 3-ethylaniline
SMILES CCC1=CC=CC(N)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -8°C
Color Beige to White or Gray
Density 0.9700g/mL
Boiling Point 212°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula C2H5C6H4NH2
Refractive Index 1.5540 to 1.556
Quantity 250 g
Beilstein 12, 1090
Specific Gravity 0.97
Formula Weight 121.18
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Ethylaniline
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Ethylaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.