Learn More
3-Ethylaniline, 98%, Thermo Scientific Chemicals
£44.20 - £723.00
Chemical Identifiers
CAS | 587-02-0 |
---|---|
Molecular Formula | C8H11N |
Molecular Weight (g/mol) | 121.18 |
MDL Number | MFCD00007818 |
InChI Key | AMKPQMFZCBTTAT-UHFFFAOYSA-N |
Synonym | m-ethylaniline, benzenamine, 3-ethyl, 3-ethylbenzenamine, 3-ethylphenylamine, aniline, m-ethyl, unii-w5u3oqy77o, aniline, 3-ethyl, w5u3oqy77o, methylanilin, 3-ethyl aniline |
PubChem CID | 11475 |
IUPAC Name | 3-ethylaniline |
SMILES | CCC1=CC=CC(N)=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10753561
|
Thermo Scientific Acros
174550100 |
10 g | Glass bottle |
£44.20
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10760681
|
Thermo Scientific Acros
174550500 |
50 g | Glass bottle |
£143.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10360954
|
Thermo Scientific Acros
174552500 |
250 g | Glass bottle |
£723.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
587-02-0 | |
121.18 | |
AMKPQMFZCBTTAT-UHFFFAOYSA-N | |
11475 | |
CCC1=CC=CC(N)=C1 |
C8H11N | |
MFCD00007818 | |
m-ethylaniline, benzenamine, 3-ethyl, 3-ethylbenzenamine, 3-ethylphenylamine, aniline, m-ethyl, unii-w5u3oqy77o, aniline, 3-ethyl, w5u3oqy77o, methylanilin, 3-ethyl aniline | |
3-ethylaniline |
Specifications
587-02-0 | |
100.0 | |
Beige to White or Gray | |
212°C | |
Authentic | |
C8H11N | |
1.5540 to 1.556 | |
12, 1090 | |
m-ethylaniline, benzenamine, 3-ethyl, 3-ethylbenzenamine, 3-ethylphenylamine, aniline, m-ethyl, unii-w5u3oqy77o, aniline, 3-ethyl, w5u3oqy77o, methylanilin, 3-ethyl aniline | |
CCC1=CC=CC(N)=C1 | |
121.18 | |
121.18 | |
Powder |
97.5 | |
-8°C | |
0.9700g/mL | |
85°C | |
97% min. (GC) | |
C2H5C6H4NH2 | |
MFCD00007818 | |
0.97 | |
AMKPQMFZCBTTAT-UHFFFAOYSA-N | |
3-ethylaniline | |
11475 | |
97% | |
3-Ethylaniline |
Safety and Handling
GHS H Statement:
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 209-594-8
- 3-Ethylaniline
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H373-May cause damage to organs.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only