missing translation for 'onlineSavingsMsg'
Learn More

3-Ethylamino-4-methylphenol, tech. 90%

Product Code. 11343366
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11343366 25 g 25g
11333366 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11343366 Supplier Thermo Scientific Alfa Aesar Supplier No. L12768.14
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 120-37-6 | C9H13NO | 151.21 g/mol

3-Ethylamino-4-methylphenol was used in the synthesis of rhodol by undergoing condensation reaction with 2-(2,4-dihydroxybenzoyl)benzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethylamino-4-methylphenol was used in the synthesis of rhodol by undergoing condensation reaction with 2-(2,4-dihydroxybenzoyl)benzoic acid.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-37-6
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00002382
InChI Key CTGSQPRDMHCIMM-UHFFFAOYSA-N
Synonym 3-ethylamino-4-methylphenol, 3-ethylamino-p-cresol, phenol, 3-ethylamino-4-methyl, ccris 4639, 3-ethylamino-4-methyl-phenol, unii-8002qme89h, 3-ethylamino-4-cresol, 3-ethylamino p-cresol, dsstox_cid_5269
PubChem CID 8428
IUPAC Name 3-(ethylamino)-4-methylphenol
SMILES CCNC1=CC(O)=CC=C1C

Specifications

Melting Point 84°C to 92°C
Boiling Point 135°C to 140°C (3 mmHg)
Quantity 25 g
Beilstein 3241548
Formula Weight 151.21
Percent Purity 90%
Grade Technical
Chemical Name or Material 3-Ethylamino-4-methylphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.