missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-5-(2-hydroxyethyl)-4-methylthiazolium bromide, 98%

Product Code. 11916781 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11916781 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11916781 Supplier Thermo Scientific Acros Supplier No. 437760250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 54016-70-5 | C8H14BrNOS | 252.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 54016-70-5
CAS Min % 97.5
CAS Max % 100.0
Melting Point 85°C to 88°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5% (Argentometry)
Packaging Glass bottle
Molecular Formula C8H14BrNOS
Quantity 25 g
Synonym 3-ethyl-5-2-hydroxyethyl-4-methylthiazolium bromide, 3-ethyl-5-2-hydroxyethyl-4-methylthiazoliumbromide, 3-ethyl-5-2-hydroxyethyl-4-methylthiazol-3-ium bromide, thiazolium, 3-ethyl-5-2-hydroxyethyl-4-methyl-, bromide, 2-3-ethyl-4-methyl-thiazol-3-ium-5-yl ethanol bromide, 3-ethyl-5-2-hydroxyethyl-4-methyl-1,3-thiazol-3-ium bromide, pubchem15236, acmc-1area, ksc272q3h, 3ethyl-5-2-hydroxyethyl-4-methylthiazolium bromide
InChI Key BDQRQMLWZJQQKS-UHFFFAOYSA-M
SMILES CC[N+]1=CSC(=C1C)CCO.[Br-]
IUPAC Name 2-(3-ethyl-4-methyl-1,3-thiazol-3-ium-5-yl)ethanol;bromide
Molecular Weight (g/mol) 252.17
PubChem CID 9964923
Formula Weight 252.17
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3-Ethyl-5-(2-hydroxyethyl)-4-methylthiazolium bromide
Show More Show Less
compliance-icons
Product Identifier
  • 3-Ethyl-5-(2-hydroxyethyl)-4-methylthiazolium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.