missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-3-oxetanemethanol, 96%, Thermo Scientific™

Product Code. 10108802 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10108802 5 g Glass bottle 5g
10130613 1 g Glass bottle 1g
10668034 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10108802 Supplier Thermo Scientific Acros Supplier No. 377700050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 3047-32-3
Molecular Weight (g/mol) 116.16
MDL Number MFCD00190143
InChI Key UNMJLQGKEDTEKJ-UHFFFAOYSA-N
Synonym 3-ethyl-3-oxetanemethanol, 3-ethyloxetan-3-yl methanol, 3-oxetanemethanol, 3-ethyl, 3-ethyloxetane-3-methanol, unii-622325vbyo, 3-ethyl-3-hydroxymethyloxetane, 3-hydroxymethyl-3-ethyloxetane, 3-ethyl-3-hydroxymethyl oxetane, 3-ethyl-oxetan-3-yl-methanol, 2-hydroxymethyl-2-ethyl-1,3-epoxypropane
PubChem CID 76444
IUPAC Name (3-ethyloxetan-3-yl)methanol
SMILES CCC1(COC1)CO

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless
Density 1.0190g/mL
Boiling Point 96.0°C (4.0 mmHg)
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C6H12O2
Refractive Index 1.4520 to 1.4540
Quantity 5 g
Specific Gravity 1.019
Formula Weight 116.16
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3-Ethyl-3-oxetanemethanol
Show More Show Less
compliance-icons
Product Identifier
  • 3-Ethyl-3-oxetanemethanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.