missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-2,5-dihydro-4-methyl-2-oxo-N-(2-phenylethyl)-1H-pyrrole-1-carboxamide, TRC

Product Code. 30387814
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30387814 50 mg 50mg
30383174 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30387814 Supplier Toronto Research Chemicals Supplier No. TRCE91653550MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 247098-18-6
Molecular Formula C16 H20 N2 O2
Molecular Weight (g/mol) 272.34
Synonym 1H-Pyrrole-1-carboxamide, 3-ethyl-2,5-dihydro-4-methyl-2-oxo-N-(2-phenylethyl)-, 3-Ethyl-2,5-dihydro-4-methyl-2-oxo-N-(2-phenylethyl)-1H-pyrrole-1-carboxamide, 3-Ethyl-4-methyl-2-oxo-N-(2-phenylethyl)-2,5-dihydro-1H-pyrrole-1-carboxamide
IUPAC Name 4-ethyl-3-methyl-5-oxo-N-(2-phenylethyl)-2H-pyrrole-1-carboxamide
SMILES CCC1=C(C)CN(C(=O)NCCc2ccccc2)C1=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C16H20N2O2/c1-3-14-12(2)11-18(15(14)19)16(20)17-10-9-13-7-5-4-6-8-13/h4-8H,3,9-11H2,1-2H3,(H,17,20)
Formula Weight 272.1525
Chemical Name or Material 3-Ethyl-2,5-dihydro-4-methyl-2-oxo-N-(2-phenylethyl)-1H-pyrr
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.