missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-2,5-dihydro-4-methyl-2-oxo-N-(2-phenylethyl)-1H-pyrrole-1-carboxamide, TRC

Product Code. 30383174
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383174 500 mg -
30387814 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30383174 Supplier Toronto Research Chemicals Supplier No. TRCE916535500MG
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 247098-18-6
Molecular Formula C16 H20 N2 O2
Molecular Weight (g/mol) 272.34
Synonym 1H-Pyrrole-1-carboxamide, 3-ethyl-2,5-dihydro-4-methyl-2-oxo-N-(2-phenylethyl)-, 3-Ethyl-2,5-dihydro-4-methyl-2-oxo-N-(2-phenylethyl)-1H-pyrrole-1-carboxamide, 3-Ethyl-4-methyl-2-oxo-N-(2-phenylethyl)-2,5-dihydro-1H-pyrrole-1-carboxamide
IUPAC Name 4-ethyl-3-methyl-5-oxo-N-(2-phenylethyl)-2H-pyrrole-1-carboxamide
SMILES CCC1=C(C)CN(C(=O)NCCc2ccccc2)C1=O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C16H20N2O2/c1-3-14-12(2)11-18(15(14)19)16(20)17-10-9-13-7-5-4-6-8-13/h4-8H,3,9-11H2,1-2H3,(H,17,20)
Formula Weight 272.1525
Chemical Name or Material 3-Ethyl-2,5-dihydro-4-methyl-2-oxo-N-(2-phenylethyl)-1H-pyrr
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.